Benzenesulfonyl fluoride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04689-03
L04689-06
CAAAAL04689-03EA
37.29
CAD
CAAAAL04689-03
CAAAAL04689-06
Benzenesulfonyl fluoride 98%
Benzenesulphonyl fluoride
Fieser: 11,40
Purity: 98%
CAS No.: 368-43-4
Molecular Formula: C6H5FO2S
Formula Weight: 160.17
Flash Point: 87°(188°F)
Refractive Index: 1.4920
Boiling Point: 203-204°
Beilstein Reference: 2206835
UN No.: 2922
MDL No.: MFCD00007412
Formula:
C₆H₅FO₂S MW: 160.17 g/mol Boiling Pt: 203…204 °C Density: 1.334 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007412 CAS Number: 368-43-4 EINECS: 206-707-2 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...