DL-Mandelamide 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Hydroxy-2-phenylacetamide
L04679-03
L04679-06
CAAAAL04679-03EA
74.73
CAD
CAAAAL04679-03
CAAAAL04679-06
DL-Mandelamide 97%
DL-Mandelamide
Purity: 97%
CAS No.: 4410-31-5
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: 132-135°
Beilstein Reference: 2208677
MDL No.: MFCD00025495
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Melting Pt: 132…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025495 CAS Number: 4410-31-5 EINECS: 626-245-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...