Ethyl-2-oxocyclopentanecarboxylate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylcyclopentanone-2-carboxylate, Ethyl 2-oxocyclopentanecarboxylate

L04528-14 L04528-22
CAAAAL04528-14EA 68.26 CAD
CAAAAL04528-14 CAAAAL04528-22
Ethyl-2-oxocyclopentanecarboxylate 97+%
Ethyl 2-oxocyclopentanecarboxylate

Purity: 97+% CAS No.: 611-10-9 Molecular Formula: C8H12O3 Formula Weight: 156.18 Flash Point: 77°(170°F) Refractive Index: 1.4520 Boiling Point: 225-228° Beilstein Reference: 387787 MDL No.: MFCD00001412

Formula: C₈H₁₂O₃
Boiling Pt: 225…228 °C
Density: 1,079
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001412
CAS Number: 611-10-9
EINECS: 210-253-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR