Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Benzyloxyindole-2-carboxylic acid ethyl ester, Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate

L04440-03 L04440-06
CAAAAL04440-03EA 113.13 CAD
CAAAAL04440-03 CAAAAL04440-06
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 97%
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate

Purity: 97% CAS No.: 37033-95-7 Molecular Formula: C18H17NO3 Formula Weight: 295.34 Storage Temperature: -30°C to -10°C Melting Point: 160-165° Beilstein Reference: 281129 MDL No.: MFCD00022702

Formula: C₁₈H₁₇NO₃
MW: 295.34 g/mol
Melting Pt: 160…165 °C
Storage Temperature: Freezer
MDL Number: MFCD00022702
CAS Number: 37033-95-7
EINECS: 253-322-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR