Methyl adipoyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04377-04
L04377-09
L04377-18
CAAAAL04377-04EA
52.3
CAD
CAAAAL04377-04
CAAAAL04377-09
CAAAAL04377-18
Methyl adipoyl chloride 96%
Methyl adipoyl chloride
Purity: 96%
CAS No.: 35444-44-1
Molecular Formula: C7H11ClO3
Formula Weight: 178.62
Refractive Index: 1.4470
Boiling Point: 76°/0.8mm
Beilstein Reference: 1766187
UN No.: 3265
MDL No.: MFCD00013661
Formula:
C₇H₁₁ClO₃ Boiling Pt: 76 °C/0.8 mm Density: 1.050 Storage Temperature: Ambient |
MDL Number:
MFCD00013661 CAS Number: 35444-44-1 EINECS: 252-569-1 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...