Methoxycarbonylsulfenyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
S-Chloro O-Methyl Thiocarbonate
L04350-03
L04350-06
CAAAAL04350-03EA
35.71
CAD
CAAAAL04350-03
CAAAAL04350-06
Methoxycarbonylsulfenyl chloride 95%
Methoxycarbonylsulfenyl chloride
Purity: 95%
CAS No.: 26555-40-8
Molecular Formula: C2H3ClO2S
Formula Weight: 126.56
Storage Temperature: 2°C to 8°C
Flash Point: 54°(129°F)
Refractive Index: 1.4820
Boiling Point: 73-74°/100mm
Beilstein Reference: 1849908
UN No.: 2920
MDL No.: MFCD00013648
Formula:
C₂H₃ClO₂S MW: 126.55 g/mol Boiling Pt: 73…74 °C/100 mm Density: 1.398 Flash Pt: 54 °C(129 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013648 CAS Number: 26555-40-8 EINECS: 247-801-3 UN: 2920 ADR: "8",II |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...