2-Amino-4'-bromoacetophenone hydrochloride 98%
Supplier: Thermo Scientific Chemicals
L04292-04
L04292-09
CAAAAL04292-04EA
99.7
CAD
CAAAAL04292-04
CAAAAL04292-09
2-Amino-4'-bromoacetophenone hydrochloride 98%
2-Amino-4'-bromoacetophenone hydrochloride
Purity: 98%
CAS No.: 5467-72-1
Molecular Formula: C8H8BrNO.HCl
Formula Weight: 250.53
Melting Point: 280-283°
Beilstein Reference: 3630035
Formula:
C₈H₉BrClNO Melting Pt: 280…283 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051998 CAS Number: 5467-72-1 EINECS: 226-778-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...