Trimethylsilylmethanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04251-03
L04251-06
CAAAAL04251-03EA
72.52
CAD
CAAAAL04251-03
CAAAAL04251-06
Trimethylsilylmethanol 95%
Trimethylsilylmethanol
Fieser: 14,331
Purity: 95%
CAS No.: 3219-63-4
Molecular Formula: (CH3)3SiCH2OH
Formula Weight: 104.23
Flash Point: 31°(87°F)
Refractive Index: 1.4190
Boiling Point: 121-122°
Beilstein Reference: 1731486
UN No.: 1993
MDL No.: MFCD00004596
Formula:
C₄H₁₂OSi Boiling Pt: 121…122 °C Density: 0.827 Flash Pt: 31 °C(87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004596 CAS Number: 3219-63-4 EINECS: 221-746-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...