4'-Methoxy-2'-nitroacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Acetamido-3-Nitroanisole, N-(4-Methoxy-2-nitro-phenyl)-acetamide, 4-Methoxy-2-nitroacetanilide, N-(4-Methoxy-2-nitrophenyl)acetamide

L04218-06 L04218-14
CAAAAL04218-06EA 61.15 CAD
CAAAAL04218-06 CAAAAL04218-14
4'-Methoxy-2'-nitroacetanilide 98+%
4'-Methoxy-2'-nitroacetanilide

Purity: 98+% CAS No.: 119-81-3 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 Melting Point: 117-118° Beilstein Reference: 2809678

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Melting Pt: 117…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00017018
CAS Number: 119-81-3
EINECS: 204-353-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR