Polygalacturonic acid, M.W. 25,000-50,000
Supplier: Thermo Scientific Chemicals
L04206-14
L04206-22
L04206-36
CAAAAL04206-14EA
88.16
CAD
CAAAAL04206-14
CAAAAL04206-22
CAAAAL04206-36
Polygalacturonic acid, M.W. 25,000-50,000
Polygalacturonic acid
CAS No.: 25990-10-7
Optical Rotation: +250° (c=1 in 0.1N NaOH)
MDL No.: MFCD00131972
Storage Temperature:
Ambient |
MDL Number:
MFCD00131972 CAS Number: 25990-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...