4-Chloro-2,5-dimethoxyaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,5-Dimethoxy-4-chloroaniline
L04202-14
L04202-22
CAAAAL04202-14EA
49.61
CAD
CAAAAL04202-14
CAAAAL04202-22
4-Chloro-2,5-dimethoxyaniline 98%
4-Chloro-2,5-dimethoxyaniline
Purity: 98%
CAS No.: 6358-64-1
Molecular Formula: C8H10ClNO2
Formula Weight: 187.63
Flash Point: 163°(325°F)
Melting Point: 118-121°
Beilstein Reference: 880445
MDL No.: MFCD00014893
Formula:
C₈H₁₀ClNO₂ MW: 187.63 g/mol Melting Pt: 118…121 °C Flash Pt: 163 °C(325 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014893 CAS Number: 6358-64-1 EINECS: 228-782-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...