D(+)-α-Acetobromoglucose 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with calcium carbonate 2%
L04151-09
L04151-18
CAAAAL04151-09EA
184.86
CAD
CAAAAL04151-09
CAAAAL04151-18
D(+)-α-Acetobromoglucose 97% stabilized
D(+)-α-Acetobromoglucose
Purity: 97%
CAS No.: 572-09-8
Molecular Formula: C14H19BrO9
Formula Weight: 411.22
Storage Temperature: -30°C to -10°C
Melting Point: 84-89°
Beilstein Reference: 96669
Merck Reference: 14,60
Optical Rotation: +198° (c=3 in chloroform)
MDL No.: MFCD00063254
Formula:
C₁₄H₁₉BrO₉ Melting Pt: 84…89 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063254 CAS Number: 572-09-8 EINECS: 209-339-0 Merck Index: 14,60 |
Specification Test Results
Optical rotation | +193° (c=3 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...