Ethyl hydrogen sebacate 98%

Supplier: Thermo Scientific Chemicals
Warning

L04134-06 L04134-14
CAAAAL04134-06EA 169.06 CAD
CAAAAL04134-06 CAAAAL04134-14
Ethyl hydrogen sebacate 98%
Ethyl hydrogen sebacate

Purity: 98% CAS No.: 693-55-0 Molecular Formula: CH3CH2O2C(CH2)8CO2H Formula Weight: 230.30 Melting Point: 35-38° Beilstein Reference: 1782569 MDL No.: MFCD00014387

Formula: CH₃CH₂O₂C(CH₂)₈CO₂H
MW: 230.30 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00014387
CAS Number: 693-55-0
EINECS: 211-753-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR