N-Methyl-N-trimethylsilylacetamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04101-04
L04101-09
CAAAAL04101-04EA
51.03
CAD
CAAAAL04101-04
CAAAAL04101-09
N-Methyl-N-trimethylsilylacetamide 98%
N-Methyl-N-trimethylsilylacetamide
Purity: 98%
CAS No.: 7449-74-3
Molecular Formula: C6H15NOSi
Formula Weight: 145.28
Flash Point: 48°(118°F)
Refractive Index: 1.4390
Boiling Point: 48-49°/11mm
Beilstein Reference: 506975
UN No.: 1993
MDL No.: MFCD00040758
Formula:
C₆H₁₅NOSi Boiling Pt: 48…49 °C/11 mm Density: 0.906 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040758 CAS Number: 7449-74-3 EINECS: 231-217-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...