1,2-Bis(phenylsulfonyl)ethane ≥98%
Supplier: Thermo Scientific Chemicals
L04083-06
L04083-14
CAAAAL04083-06EA
118.03
CAD
CAAAAL04083-06
CAAAAL04083-14
1,2-Bis(phenylsulfonyl)ethane ≥98%
1,2-Bis(phenylsulfonyl)ethane
Purity: 98+%
CAS No.: 599-94-0
Molecular Formula: C14H14O4S2
Formula Weight: 310.39
Melting Point: 179-180°
Beilstein Reference: 1888062
MDL No.: MFCD00041256
Formula:
C₁₄H₁₄O₄S₂ Melting Pt: 179…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041256 CAS Number: 599-94-0 EINECS: 209-979-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...