4-(2-Chloroethyl)benzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04068-09
L04068-18
CAAAAL04068-09EA
161.16
CAD
CAAAAL04068-09
CAAAAL04068-18
4-(2-Chloroethyl)benzoic acid 97%
4-(2-Chloroethyl)benzoic acid
Purity: 97%
CAS No.: 20849-78-9
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Melting Point: 187-189°
Beilstein Reference: 2084041
MDL No.: MFCD00013995
Formula:
C₉H₉ClO₂ Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013995 CAS Number: 20849-78-9 EINECS: 244-080-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...