7-Ethoxy-7-oxoheptanoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monoethyl pimelate, Pimelic acid monoethyl ester, Ethyl yydrogen pimelate

L04025-06 L04025-14
CAAAAL04025-06EA 165.9 CAD
CAAAAL04025-06 CAAAAL04025-14
7-Ethoxy-7-oxoheptanoic acid 98%
7-Ethoxy-7-oxoheptanoic acid

Purity: 98% CAS No.: 33018-91-6 Molecular Formula: C9H16O4 Formula Weight: 188.22 Flash Point: 110°(230°F) Refractive Index: 1.4420 Melting Point: 10° Boiling Point: 135-138°/1mm Beilstein Reference: 1774583 MDL No.: MFCD00801202

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 135…138 °C/1 mm
Melting Pt: 10 °C
Density: 1.06
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014383
CAS Number: 33018-91-6
EINECS: 251-346-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR