4-Benzoylbutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04013-06
L04013-14
CAAAAL04013-06EA
82.48
CAD
CAAAAL04013-06
CAAAAL04013-14
4-Benzoylbutyric acid 97%
4-Benzoylbutyric acid
Purity: 97%
CAS No.: 1501-05-9
Molecular Formula: C11H12O3
Formula Weight: 192.21
Melting Point: 126-128°
Beilstein Reference: 641947
MDL No.: MFCD00004411
Formula:
C₁₁H₁₂O₃ Melting Pt: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004411 CAS Number: 1501-05-9 EINECS: 216-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...