5-Oxohexanoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03978-06
L03978-14
CAAAAL03978-06EA
83.42
CAD
CAAAAL03978-06
CAAAAL03978-14
5-Oxohexanoic acid 97%
5-Oxohexanoic acid
Purity: 97%
CAS No.: 3128-06-1
Molecular Formula: CH3CO(CH2)3CO2H
Formula Weight: 130.14
Flash Point:
Formula:
C₆H₁₀O₃ Boiling Pt: 274…275 °C Melting Pt: 12…14 °C Density: 1.100 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004412 CAS Number: 3128-06-1 EINECS: 221-511-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...