Ethyl phenylcyanoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

L03929-06 L03929-14
CAAAAL03929-06EA 71.26 CAD
CAAAAL03929-06 CAAAAL03929-14
Ethyl phenylcyanoacetate 98%
Ethyl phenylcyanoacetate

Purity: 98% CAS No.: 4553-07-5 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Flash Point: 110°(230°F) Refractive Index: 1.505 Boiling Point: 274-275° Beilstein Reference: 611232 MDL No.: MFCD00001866

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Boiling Pt: 274…275 °C
Density: 1.090
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001866
CAS Number: 4553-07-5
EINECS: 224-921-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR