3-(Trifluoromethyl)crotonic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03816-06
L03816-14
CAAAAL03816-06EA
143.62
CAD
CAAAAL03816-06
CAAAAL03816-14
3-(Trifluoromethyl)crotonic acid 96%
3-(Trifluoromethyl)crotonic acid
Purity: 96%
CAS No.: 69056-67-3
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Flash Point: 79°(174°F)
Refractive Index: 1.3850
Melting Point: 28-30°
Boiling Point: 158-163°
Beilstein Reference: 1769204
UN No.: 3261
MDL No.: MFCD00014374
Formula:
C₅H₅F₃O₂ Boiling Pt: 158…163 °C Melting Pt: 28…30 °C Density: 1.318 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014374 CAS Number: 69056-67-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...