Estradiol, Wasser_3 99% Dry weight

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: beta-Estradiol, 17β-Estradiol

L03801-03 L03801-06 L03801-14
CAAAAL03801-03EA 74.58 CAD
CAAAAL03801-03 CAAAAL03801-06 CAAAAL03801-14
Estradiol, Wasser_3 99% Dry weight
Estradiol

Purity: 99% CAS No.: 50-28-2 Molecular Formula: C18H24O2 Formula Weight: 272.39 Melting Point: 175-178° Beilstein Reference: 1914275 Merck Reference: 14,3703 Optical Rotation: +79° (c=1 in dioxane) MDL No.: MFCD00003693

Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8
Merck Index: 14,3703

Order Now

Specification Test Results

Optical rotation +79° (c=1 in dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR