Dimethyl-L-glutamate hydrochloride ≥97%
Supplier: Thermo Scientific Chemicals
L03764-06
L03764-14
CAAAAL03764-06EA
71.57
CAD
CAAAAL03764-06
CAAAAL03764-14
Dimethyl-L-glutamate hydrochloride ≥97%
Dimethyl-L-glutamate hydrochloride
Purity: 98+%
CAS No.: 23150-65-4
Molecular Formula: C7H13NO4.HCl
Formula Weight: 211.65
Storage Temperature: 2°C to 8°C
Odor: Odorless
Melting Point: 99-103°
Beilstein Reference: 4238829
Optical Rotation: +26° (c=5 in water)
MDL No.: MFCD00038879
Formula:
C₇H₁₃NO₄ Melting Pt: 99…103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038879 CAS Number: 23150-65-4 EINECS: 245-461-0 |
Specification Test Results
Optical rotation | +26° (c=5 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...