Triethylsilanol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Hydroxytriethylsilane, Triethyl(hydroxy)silane
L03738-03
L03738-06
CAAAAL03738-03EA
83.58
CAD
CAAAAL03738-03
CAAAAL03738-06
Triethylsilanol 97%
Triethylsilanol
Purity: 97%
CAS No.: 597-52-4
Molecular Formula: C6H16OSi
Formula Weight: 132.28
Storage Temperature: 2°C to 8°C
Flash Point: 57°(135°F)
Refractive Index: 1.4330
Odor: Mild
Boiling Point: 154°
Beilstein Reference: 1732848
UN No.: 1993
MDL No.: MFCD00042640
Formula:
C₆H₁₆OSi |
MDL Number:
MFCD00042640 CAS Number: 597-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...