exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride, exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride

L03620-14 L03620-22
CAAAAL03620-06EA 65.06 CAD
CAAAAL03620-06 CAAAAL03620-14 CAAAAL03620-22
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98%
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride

Purity: 98+% CAS No.: 6118-51-0 Molecular Formula: C8H6O4 Formula Weight: 166.13 Melting Point: 118-119° Beilstein Reference: 10355 MDL No.: MFCD00151506

Formula: C₈H₆O₄
MW: 166.13 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00151506
CAS Number: 6118-51-0
EINECS: 226-570-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR