N-Cyclohexylhydroxylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

L03569-03 L03569-06
CAAAAL03569-03EA 84.21 CAD
CAAAAL03569-03 CAAAAL03569-06
N-Cyclohexylhydroxylamine hydrochloride 98%
N-Cyclohexylhydroxylamine hydrochloride

Purity: 98% CAS No.: 25100-12-3 Molecular Formula: C6H13NO.HCl Formula Weight: 151.64 Melting Point: ca 166° dec. Beilstein Reference: 3672798 MDL No.: MFCD00012565

Formula: C₆H₁₃NO·HCl
Melting Pt: ∼ 166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012565
CAS Number: 25100-12-3
EINECS: 246-612-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR