N,N'-Dibenzylethylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L03540-14 L03540-22
CAAAAL03540-14EA 135.25 CAD
CAAAAL03540-14 CAAAAL03540-22
N,N'-Dibenzylethylenediamine 97%
N,N'-Dibenzylethylenediamine

Purity: 97% CAS No.: 140-28-3 Molecular Formula: C16H20N2 Formula Weight: 240.35 Flash Point: 110°(230°F) Refractive Index: 1.5650 Melting Point: 23-25° Boiling Point: 184-186°/2mm Beilstein Reference: 786668 UN No.: 2735 MDL No.: MFCD00004771

Formula: C₁₆H₂₀N₂
Boiling Pt: 184…186 °C/2 mm
Melting Pt: 23…25 °C
Density: 1.025
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004771
CAS Number: 140-28-3
EINECS: 205-408-4
UN: 3259
ADR: 8,III
Merck Index: 14,1064

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR