Cyclobutyl methyl ketone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetylcyclobutane, Ketone cyclobutyl methyl, 1-Cyclobutylethan-1-one, 1-Cyclobutylethanone, Cyclobutyl Methyl Ketone

L03521-03 L03521-06
CAAAAL03521-03EA 70.63 CAD
CAAAAL03521-03 CAAAAL03521-06
Cyclobutyl methyl ketone 97%
Cyclobutyl methyl ketone

Purity: 97% CAS No.: 3019-25-8 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 28°(82°F) Refractive Index: 1.4320 Boiling Point: 134-135° Beilstein Reference: 1924022 UN No.: 1224 MDL No.: MFCD00037148

Formula: C₆H₁₀O
Boiling Pt: 134…135 °C
Density: 0.900
Flash Pt: 28 °C(82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037148
CAS Number: 3019-25-8
EINECS: 221-163-6
UN: 1992
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR