2-Bromo-1,3-diethylbenzene 94%

Supplier: Thermo Scientific Chemicals
Warning

L03482-03 L03482-06
CAAAAL03482-03EA 99.07 CAD
CAAAAL03482-03 CAAAAL03482-06
2-Bromo-1,3-diethylbenzene 94%
2-Bromo-1,3-diethylbenzene

Purity: 94% CAS No.: 65232-57-7 Molecular Formula: C10H13Br Formula Weight: 213.12 Refractive Index: 1.5440 Boiling Point: 78-80°/2mm MDL No.: MFCD00017770

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 78…80 °C/2 mm
Density: 1,264
Storage Temperature: Ambient
MDL Number: MFCD00017770
CAS Number: 65232-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR