4,4'-Biphenyldicarbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4'-Dicyanobiphenyl, 4,4'-Dibenzonitrile, [1,1'-Biphenyl]-4,4'-dicarbonitrile, 4,4'-Bibenzonitrile
L03450-03
L03450-06
CAAAAL03450-03EA
141.88
CAD
CAAAAL03450-03
CAAAAL03450-06
4,4'-Biphenyldicarbonitrile 98%
4,4'-Biphenyldicarbonitrile
Purity: 98%
CAS No.: 1591-30-6
Molecular Formula: C14H8N2
Formula Weight: 204.23
Melting Point: 232-236°
Beilstein Reference: 1869917
UN No.: 3439
MDL No.: MFCD00013805
Formula:
C₁₄H₈N₂ MW: 204.23 g/mol Melting Pt: 232…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013805 CAS Number: 1591-30-6 EINECS: 216-468-6 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...