1-Cyclopentylethanol 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L03446-04
L03446-09
CAAAAL03446-04EA
126.72
CAD
CAAAAL03446-04
CAAAAL03446-09
1-Cyclopentylethanol 97%
1-Cyclopentylethanol
Purity: 97%
CAS No.: 52829-98-8
Molecular Formula: C7H14O
Formula Weight: 114.19
Refractive Index: 1.4580
Boiling Point: 68°/8mm
Beilstein Reference: 2069851
MDL No.: MFCD00013743
Formula:
C₇H₁₄O Boiling Pt: 68 °C/8 mm Density: 0.911 Storage Temperature: Ambient |
MDL Number:
MFCD00013743 CAS Number: 52829-98-8 EINECS: 258-208-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...