m-Toluenesulfonic acid monohydrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03419-06
L03419-14
CAAAAL03419-06EA
165.9
CAD
CAAAAL03419-06
CAAAAL03419-14
m-Toluenesulfonic acid monohydrate 97%
m-Toluenesulfonic acid monohydrate
Purity: 97%
CAS No.: 312619-56-0
Molecular Formula: C7H8O3S.H2O
Formula Weight: 190.22 (172.20anhy)
Refractive Index: 1.5270
Boiling Point: 165°/0.1mm
Beilstein Reference: 2208421
UN No.: 2586
MDL No.: MFCD00167130
Formula:
C₇H₈O₃S·H₂O Boiling Pt: 165 °C/0.1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00167130 CAS Number: 312619-56-0 UN: 2586 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...