Trimethylsilyl isothiocyanate 94%

Supplier: Thermo Scientific Chemicals
Danger

L03277-06 L03277-14
CAAAAL03277-06EA 81.37 CAD
CAAAAL03277-06 CAAAAL03277-14
Trimethylsilyl isothiocyanate 94%
Trimethylsilyl isothiocyanate

Purity: 94% CAS No.: 2290-65-5 Molecular Formula: C4H9NSSi Formula Weight: 131.27 Storage Temperature: 2°C to 8°C Flash Point: 29°(84°F) Refractive Index: 1.4820 Odor: Pungent Melting Point: -33° Boiling Point: 143-144° Beilstein Reference: 1745262 UN No.: 1992 MDL No.: MFCD00004797

Formula: C₄H₉NSSi
MW: 131.27 g/mol
Boiling Pt: 143…144 °C
Melting Pt: –33 °C
Density: 0.932
Flash Pt: 29 °C (84 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004797
CAS Number: 2290-65-5
EINECS: 218-929-7
UN: 1992
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR