Ethyl-2-methylnicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03256-06
L03256-14
CAAAAL03256-03EA
33.65
CAD
CAAAAL03256-03
CAAAAL03256-06
CAAAAL03256-14
Ethyl-2-methylnicotinate 97%
Ethyl 2-methylnicotinate
Purity: 97%
CAS No.: 1721-26-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 102°(215°F)
Refractive Index: 1.5050
Odor: Recognizable
Boiling Point: 107-108°/13mm
Beilstein Reference: 118200
MDL No.: MFCD00006336
Formula:
C₉H₁₁NO₂ Boiling Pt: 107…108 °C/13 mm Density: 1,070 Flash Pt: 102 °C(215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006336 CAS Number: 1721-26-2 EINECS: 217-013-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...