(1R)-endo-(+)-Fenchyl alcohol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03211-22
L03211-36
CAAAAL03211-22EA
54.67
CAD
CAAAAL03211-22
CAAAAL03211-36
(1R)-endo-(+)-Fenchyl alcohol 96%
(1R)-endo-(+)-Fenchyl alcohol
Purity: 96%
CAS No.: 2217-02-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 73°(163°F)
Odor: Camphor-like
Melting Point: 37-44°
Boiling Point: 201-202°
Beilstein Reference: 1850917
Optical Rotation: +10.5° (c=10 in ethanol)
MDL No.: MFCD00003760
Formula:
C₁₀H₁₈O Boiling Pt: 201…202 °C Melting Pt: 37…44 °C Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003760 CAS Number: 2217-02-9 EINECS: 216-639-5 |
Specification Test Results
Optical rotation | +10.5° (c=10 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...