N'-Ethyl-N,N-dimethylethylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N-dimethyl-N-ethylethylenediamine

L03180-06 L03180-14
CAAAAL03180-06EA 69.99 CAD
CAAAAL03180-06 CAAAAL03180-14
N'-Ethyl-N,N-dimethylethylenediamine 98%
N'-Ethyl-N,N-dimethylethylenediamine

Purity: 98% CAS No.: 123-83-1 Molecular Formula: CH3CH2NHCH2CH2N(CH3)2 Formula Weight: 116.21 Flash Point: 23°(73°F) Refractive Index: 1.4225 Odor: Amine-like Boiling Point: 134-135° Beilstein Reference: 773675 UN No.: 2734 MDL No.: MFCD00009034

Formula: CH₃CH₂NHCH₂CH₂N(CH₃)₂
MW: 116.21 g/mol
Boiling Pt: 134…135 °C
Density: 0.818
Flash Pt: 23 °C(73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009034
CAS Number: 123-83-1
EINECS: 204-656-0
UN: 2734
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR