Nordihydroguaiaretic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03149-03
L03149-06
CAAAAL03149-03EA
120.71
CAD
CAAAAL03149-03
CAAAAL03149-06
Nordihydroguaiaretic acid 97%
Nordihydroguaiaretic acid
Naturally occuring antioxidant and inhibitor of lipoxygenase
Purity: 97%
CAS No.: 500-38-9
Molecular Formula: C18H22O4
Formula Weight: 302.37
Odor: Odorless
Melting Point: 184-188°
Beilstein Reference: 2056825
Merck Reference: 14,6693
MDL No.: MFCD00002206
Formula:
C₁₈H₂₂O₄ Melting Pt: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002206 CAS Number: 500-38-9 EINECS: 207-903-0 Merck Index: 14,6693 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...