Phenyl-p-tolylsulfone ≥98%
Supplier: Thermo Scientific Chemicals
L03128-09
L03128-18
CAAAAL03128-09EA
118.18
CAD
CAAAAL03128-09
CAAAAL03128-18
Phenyl-p-tolylsulfone ≥98%
Phenyl-p-tolylsulfone
Purity: 98+%
CAS No.: 640-57-3
Molecular Formula: C13H12O2S
Formula Weight: 232.30
Odor: Odorless
Melting Point: 125-129°
Beilstein Reference: 1872872
MDL No.: MFCD00025038
Formula:
C₁₃H₁₂O₂S Melting Pt: 125…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025038 CAS Number: 640-57-3 EINECS: 211-364-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...