Pentamethylene sulfide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03063-03
L03063-06
CAAAAL03063-03EA
49.93
CAD
CAAAAL03063-03
CAAAAL03063-06
Pentamethylene sulfide 98%
Pentamethylene sulfide
Purity: 98%
CAS No.: 1613-51-0
Molecular Formula: C5H10S
Formula Weight: 102.20
Flash Point: 21°(69°F)
Refractive Index: 1.5060
Odor: Unpleasant
Boiling Point: 140-142°
Beilstein Reference: 102437
UN No.: 1993
MDL No.: MFCD00006662
Formula:
C₅H₁₀S Boiling Pt: 140…142 °C Density: 0.988 Flash Pt: 21 °C(69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006662 CAS Number: 1613-51-0 EINECS: 216-561-1 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...