Norcantharidin ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02947-06
L02947-14
CAAAAL02947-06EA
118.34
CAD
CAAAAL02947-06
CAAAAL02947-14
Norcantharidin ≥98%
Norcantharidin
Purity: 98+%
CAS No.: 29745-04-8
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 111-116°
Beilstein Reference: 84287
MDL No.: MFCD00213361
Formula:
C₈H₈O₄ Melting Pt: 111…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213361 CAS Number: 29745-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...