4-(Methylthio)toluene 97%
Supplier: Thermo Scientific Chemicals
Warning
|
L02933-06
L02933-14
CAAAAL02933-06EA
58.46
CAD
CAAAAL02933-06
CAAAAL02933-14
4-(Methylthio)toluene 97%
4-(Methylthio)toluene
Purity: 97%
CAS No.: 623-13-2
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 85°(185°F)
Refractive Index: 1.5730
Odor: Unpleasant
Boiling Point: 81-82°/18mm
Beilstein Reference: 1905733
UN No.: 3334
MDL No.: MFCD00008560
Formula:
C₈H₁₀S Boiling Pt: 81…82 °C/18 mm Density: 1.029 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008560 CAS Number: 623-13-2 EINECS: 210-773-8 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...