Cholesterol pelargonate
Supplier: Thermo Scientific Chemicals
L02857-14
L02857-22
CAAAAL02857-14EA
88.48
CAD
CAAAAL02857-14
CAAAAL02857-22
Cholesterol pelargonate
Cholesterol pelargonate
CAS No.: 1182-66-7
Molecular Formula: C36H62O2
Formula Weight: 526.89
Odor: Odorless
Melting Point: 75-79°
Beilstein Reference: 3179820
Optical Rotation: -30° (c=5 in chloroform)
MDL No.: MFCD00003643
Formula:
C₃₆H₆₂O₂ Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003643 CAS Number: 1182-66-7 EINECS: 214-658-3 |
Specification Test Results
Optical rotation | -30° (c=5 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...