N,N-Dimethylformamide dineopentyl acetal 98%

Supplier: Thermo Scientific Chemicals
Warning

L02728-09 L02728-18
CAAAAL02728-09EA 131.77 CAD
CAAAAL02728-09 CAAAAL02728-18
N,N-Dimethylformamide dineopentyl acetal 98%
N,N-Dimethylformamide dineopentyl acetal

Fieser: 1,283 6,222 9,183 12,204


Purity: 98% CAS No.: 4909-78-8 Molecular Formula: C13H29NO2 Formula Weight: 231.38 Flash Point: 63°(145°F) Refractive Index: 1.4117 Boiling Point: 85-87°/10mm Beilstein Reference: 741992 MDL No.: MFCD00008851

Formula: C₁₃H₂₉NO₂
MW: 231.38 g/mol
Boiling Pt: 85…87 °C/10 mm
Density: 0.830
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008851
CAS Number: 4909-78-8
EINECS: 225-536-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR