4-Hydroxy-3,5-diiodobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02595-09
L02595-18
CAAAAL02595-09EA
126.24
CAD
CAAAAL02595-09
CAAAAL02595-18
4-Hydroxy-3,5-diiodobenzoic acid 97%
4-Hydroxy-3,5-diiodobenzoic acid
Purity: 97%
CAS No.: 618-76-8
Molecular Formula: C7H4I2O3
Formula Weight: 389.91
Melting Point: ca 275° dec.
Beilstein Reference: 2212145
MDL No.: MFCD00016532
Formula:
C₇H₄I₂O₃ Melting Pt: ca 275 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016532 CAS Number: 618-76-8 EINECS: 210-562-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...