N-Methyliminodiacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2'-(Methylazanediyl)diacetic acid
L02589-06
L02589-14
CAAAAL02589-06EA
89.74
CAD
CAAAAL02589-06
CAAAAL02589-14
N-Methyliminodiacetic acid 99%
N-Methyliminodiacetic acid
Purity: 99%
CAS No.: 4408-64-4
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Melting Point: ca 220° dec.
Beilstein Reference: 1765449
Formula:
C₅H₉NO₄ Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004284 CAS Number: 4408-64-4 EINECS: 224-557-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...