1,2-Bis(chlorodimethylsilyl)ethane 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02570-14
L02570-22
CAAAAL02570-14EA
103.17
CAD
CAAAAL02570-14
CAAAAL02570-22
CAAAAL02570-36
1,2-Bis(chlorodimethylsilyl)ethane 90%, tech.
1,2-Bis(chlorodimethylsilyl)ethane
Purity: 90%
CAS No.: 13528-93-3
Molecular Formula: C6H16Cl2Si2
Formula Weight: 215.27
Flash Point: 65°(149°F)
Melting Point: 34-40°
Boiling Point: 198-200°
Beilstein Reference: 1920141
UN No.: 3261
MDL No.: MFCD00000505
Formula:
C₆H₁₆Cl₂Si₂ Boiling Pt: 198…200 °C Melting Pt: 34…40 °C Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000505 CAS Number: 13528-93-3 EINECS: 236-871-0 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...