Diphenylpropylphosphine 97%

Supplier: Thermo Scientific Chemicals
Warning

L02402-03 L02402-06 L02402-14
CAAAAL02402-03EA 61.3 CAD
CAAAAL02402-03 CAAAAL02402-06 CAAAAL02402-14
Diphenylpropylphosphine 97%
Diphenylpropylphosphine

Purity: 97% CAS No.: 7650-84-2 Molecular Formula: C15H17P Formula Weight: 228.28 Flash Point: 110°(230°F) Refractive Index: 1.6035 Odor: Unpleasant Boiling Point: 163-164°/14mm Beilstein Reference: 642795 MDL No.: MFCD00015216

Formula: C₁₅H₁₇P
Boiling Pt: 163…164 °C/14 mm
Density: 1.008
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015216
CAS Number: 7650-84-2
EINECS: 231-607-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR