Dimethyl allylmalonate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl 2-allylmalonate

L02376-09 L02376-18
CAAAAL02376-09EA 77.74 CAD
CAAAAL02376-09 CAAAAL02376-18
Dimethyl allylmalonate 97%
Dimethyl allylmalonate

Purity: 97% CAS No.: 40637-56-7 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 95°(203°F) Refractive Index: 1.4350 Melting Point: -80° Boiling Point: 207° Beilstein Reference: 1772822 MDL No.: MFCD00014959

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 207 °C
Melting Pt: < –80 °C
Density: 1.074
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014959
CAS Number: 40637-56-7
EINECS: 255-017-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR