1-Adamantanecarbonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Adamantanecarboxylic acid chloride
L02352-09
L02352-18
CAAAAL02352-09EA
140.15
CAD
CAAAAL02352-09
CAAAAL02352-18
1-Adamantanecarbonyl chloride 97%
1-Adamantanecarbonyl chloride
Fieser: 2,15
Purity: 97%
CAS No.: 2094-72-6
Molecular Formula: C11H15ClO
Formula Weight: 198.69
Melting Point: 48-53°
Boiling Point: 135-136°/10mm
Beilstein Reference: 389960
UN No.: 3261
MDL No.: MFCD00074724
Formula:
C₁₁H₁₅ClO Boiling Pt: 135…136 °C/10 mm Melting Pt: 48…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074724 CAS Number: 2094-72-6 EINECS: 218-252-7 UN: 1759 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...