2-Hydroxy-n-octanoic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02314-03
L02314-06
CAAAAL02314-03EA
59.41
CAD
CAAAAL02314-03
CAAAAL02314-06
2-Hydroxy-n-octanoic acid ≥98%
2-Hydroxy-n-octanoic acid
Formula:
C₈H₁₆O₃ Melting Pt: 66…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014410 CAS Number: 617-73-2 EINECS: 210-524-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...